siRNA Details
Gene Symbol: ATAD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317238.1 | NP_001304167.1 | 13, 14 | 1869 | ||
NM_031921.5 | NP_114127.3 | 15, 16 | 1752 | |||
XM_011542243.2 | XP_011540545.1 | 14, 15 | 1488 | |||
XM_011542244.1 | XP_011540546.1 | 15, 16 | 1919 | |||
GenBank mRNA | AB033099.1 | - | 13, 14 | 1958 | ||
AK024028.1 | - | 13, 14 | 1337 | |||
AK127857.1 | - | 9, 10 | 2523 | |||
AK290586.1 | - | 15, 16 | 1744 | |||
AL834179.1 | - | 4, 5 | 2658 | |||
BC002542.2 | - | 15, 16 | 1678 | 3138578 | ||
BC018701.1 | - | 15, 16 | 1678 | 3139886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
