siRNA Details
Gene Symbol: ATAD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317238.1 | NP_001304167.1 | 14 | 2090 | ||
NM_031921.5 | NP_114127.3 | 16 | 1973 | |||
XM_011542243.2 | XP_011540545.1 | 15 | 1709 | |||
XM_011542244.1 | XP_011540546.1 | 16 | 2140 | |||
GenBank mRNA | AB033099.1 | - | 14 | 2179 | ||
AK024028.1 | - | 14 | 1558 | |||
AK127857.1 | - | 10 | 2744 | |||
AK290586.1 | - | 16 | 1965 | |||
AL834179.1 | - | 5 | 2879 | |||
BC002542.2 | - | 16 | 1899 | 3138578 | ||
BC018701.1 | - | 16 | 1899 | 3139886 |
Gene Symbol: ATAD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170535.1 | NP_001164006.1 | 16 | 1969 | ||
NM_001170536.1 | NP_001164007.1 | 16 | 1799 | |||
NM_018188.3 | NP_060658.3 | 16 | 2113 | |||
GenBank mRNA | AK001571.1 | - | 16 | 1952 | ||
AK025865.1 | - | 9 | 969 | |||
AK056099.1 | - | 16 | 1799 | |||
AK091144.1 | - | 16 | 2081 | |||
AK092833.1 | - | 1 | 2085 | |||
AK310572.1 | - | 1991 | ||||
BC007803.2 | - | 16 | 1930 | 4136170 | ||
BC011814.2 | - | 16 | 1935 | 3162989 | ||
BC014101.2 | - | 16 | 1959 | 3503438 | ||
BC033109.1 | - | 16 | 1933 | 4877449 | ||
BC063607.1 | - | 16 | 1927 | 4939339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
