siRNA Details
Gene Symbol: ACBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717528.3 | XP_006717591.3 | 1 | 1805 | ||
XM_017016884.1 | XP_016872373.1 | 1 | 1805 | |||
XM_017016885.1 | XP_016872374.1 | 1 | 1805 | |||
XM_017016886.1 | XP_016872375.1 | 1 | 1805 | |||
XM_017016890.1 | XP_016872379.1 | 1 | 1805 | |||
XM_017016893.1 | XP_016872382.1 | 1 | 1805 | |||
XM_017016894.1 | XP_016872383.1 | 1 | 1805 |
Gene Symbol: LOC107985082 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025510.1 | XP_016880999.1 | 1 | 1080 |
Gene Symbol: LRRC37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014834.4 | NP_055649.4 | 9 | 4337 | ||
XM_005257879.4 | XP_005257936.1 | 9 | 4595 | |||
XM_005257880.4 | XP_005257937.1 | 7 | 4448 | |||
XM_005257882.4 | XP_005257939.1 | 10 | 4781 | |||
XM_006722210.3 | XP_006722273.1 | 5 | 4295 | |||
XM_006722211.3 | XP_006722274.1 | 4 | 4223 | |||
XM_011525539.2 | XP_011523841.1 | 7 | 4442 | |||
XM_017025472.1 | XP_016880961.1 | 11 | 4194 | |||
XM_017025473.1 | XP_016880962.1 | 9 | 1841 | |||
GenBank mRNA | BC040501.1 | - | 9 | 4337 | 5295808 | |
CR749232.1 | - | 9 | 4338 |
Gene Symbol: LRRC37A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006607.2 | NP_001006608.2 | 9 | 4337 | ||
XM_011524841.2 | XP_011523143.1 | 8 | 4318 | |||
XM_011524842.2 | XP_011523144.1 | 9 | 4390 | |||
XM_011524843.2 | XP_011523145.1 | 7 | 4243 | |||
XM_011524844.2 | XP_011523146.1 | 7 | 4237 | |||
XM_011524846.2 | XP_011523148.1 | 6 | 4165 | |||
XM_011524848.2 | XP_011523150.1 | 5 | 4090 | |||
GenBank mRNA | AK091733.1 | - | 1 | 2249 | ||
AK126994.1 | - | 9 | 3019 | |||
AY386262.1 | - | 9 | 2758 | |||
BC088375.1 | - | 1 | 1135 | |||
BC144427.1 | - | 9 | 2000 | |||
BC144428.1 | - | 9 | 2000 |
Gene Symbol: LRRC37A4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002940.2 | 1 | 2309 | |||
GenBank mRNA | AK000982.1 | - | 1 | 438 |
Gene Symbol: LRRC37A6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003525.2 | 1 | 5437 |
Gene Symbol: LRRC37BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124001.1 | - | 1 | 495 | ||
AK124759.1 | - | 1 | 902 | |||
AL713754.1 | - | 1 | 20 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
