siRNA Details
Gene Symbol: RNF145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199380.1 | NP_001186309.1 | 10 | 1678 | ||
NM_001199381.1 | NP_001186310.1 | 10 | 1725 | |||
NM_001199382.1 | NP_001186311.1 | 10 | 1651 | |||
NM_001199383.1 | NP_001186312.1 | 10 | 1802 | |||
NM_144726.2 | NP_653327.1 | 10 | 1549 | |||
XM_017009138.1 | XP_016864627.1 | 10 | 1997 | |||
GenBank mRNA | AK056513.1 | - | 10 | 1549 | ||
AK075101.1 | - | 1 | 127 | |||
AK098802.1 | - | 5 | 750 | |||
AK292854.1 | - | 10 | 1791 | |||
AK294323.1 | - | 2 | 186 | |||
AK304228.1 | - | 10 | 1637 | |||
AK304435.1 | - | 10 | 1725 | |||
AK308394.1 | - | 10 | 1677 | |||
BC042684.1 | - | 10 | 1814 | 4826810 | ||
BC044767.1 | - | 10 | 1814 | |||
BX640806.1 | - | 3 | 408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
