siRNA Details
Gene Symbol: HSPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002157.2 | NP_002148.1 | 3, 4 | 698 | ||
GenBank mRNA | AK312104.1 | - | 3, 4 | 294 | ||
BC023518.2 | - | 3, 4 | 372 | 4304224 | ||
BG188453.1 | - | 2, 3 | 259 | |||
CN264788.1 | - | 3, 4 | 703 | |||
CR407688.1 | - | 2, 3 | 258 | |||
U07550.1 | - | 3, 4 | 299 | |||
X75821.1 | - | 2, 3 | 258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
