siRNA Details
Gene Symbol: ZNF85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256171.1 | NP_001243100.1 | 2 | 241 | ||
NM_001256172.1 | NP_001243101.1 | 2 | 241 | |||
NM_003429.4 | NP_003420.2 | 2 | 241 | |||
NR_045830.1 | 2 | 254 | ||||
XM_011528263.2 | XP_011526565.1 | 2 | 267 | |||
GenBank mRNA | BC008688.1 | - | 2 | 197 | ||
BC068066.1 | - | 2 | 218 | |||
BM755583.1 | - | 2 | 240 | |||
BP199573.1 | - | 2 | 241 | |||
DC358791.1 | - | 2 | 209 | |||
DN994243.1 | - | 2 | 228 | |||
M61866.1 | - | 2 | 205 | |||
M61868.1 | - | 1 | 60 | |||
U35376.1 | - | 2 | 211 |
Gene Symbol: ZNF208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007153.3 | NP_009084.2 | 2 | 242 | ||
XM_017027257.1 | XP_016882746.1 | 2 | 289 | |||
GenBank mRNA | CB997649.1 | - | 2 | 269 |
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278661.1 | NP_001265590.1 | 3 | 403 | ||
NM_001278662.1 | NP_001265591.1 | 3 | 383 | |||
NM_001278665.1 | NP_001265594.1 | 2 | 268 | |||
NM_001278677.1 | NP_001265606.1 | 4 | 475 | |||
NM_203282.3 | NP_975011.3 | 2 | 268 | |||
XM_011528443.1 | XP_011526745.1 | 4 | 536 | |||
XM_011528444.1 | XP_011526746.1 | 5 | 556 | |||
XM_017027514.1 | XP_016883003.1 | 4 | 466 | |||
XM_017027515.1 | XP_016883004.1 | 6 | 893 | |||
XM_017027516.1 | XP_016883005.1 | 2 | 2940 | |||
XM_017027517.1 | XP_016883006.1 | 2 | 696 | |||
GenBank mRNA | AF054180.1 | - | 2 | 254 | ||
AK291286.1 | - | 2 | 235 | |||
BC043147.1 | - | 2 | 240 | 5296940 | ||
BC044906.1 | - | 1 | 1843 | |||
BC056243.1 | - | 2 | 244 | |||
BC139749.1 | - | 2 | 199 | 40115903 | ||
BX281205.1 | - | 3 | 271 | |||
DA101675.1 | - | 4 | 393 | |||
DB286139.1 | - | 3 | 367 |
Gene Symbol: ZNF429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001415.2 | NP_001001415.2 | 2 | 230 | ||
XM_017026748.1 | XP_016882237.1 | 3 | 729 | |||
XM_017026749.1 | XP_016882238.1 | 3 | 2643 | |||
XM_017026750.1 | XP_016882239.1 | 3 | 833 | |||
XM_017026751.1 | XP_016882240.1 | 2 | 744 | |||
XM_017026752.1 | XP_016882241.1 | 2 | 286 | |||
XM_017026753.1 | XP_016882242.1 | 4 | 582 | |||
XM_017026754.1 | XP_016882243.1 | 3 | 527 | |||
XM_017026755.1 | XP_016882244.1 | 4 | 2666 | |||
XM_017026756.1 | XP_016882245.1 | 3 | 534 | |||
GenBank mRNA | AF325191.1 | - | 2 | 206 | ||
AK057495.1 | - | 2 | 243 | |||
AK308708.1 | - | 2 | 261 | |||
AY269786.1 | - | 2 | 206 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
