siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278661.1 | NP_001265590.1 | 3 | 355 | ||
NM_001278662.1 | NP_001265591.1 | 3 | 335 | |||
NM_001278665.1 | NP_001265594.1 | 2 | 220 | |||
NM_001278677.1 | NP_001265606.1 | 4 | 427 | |||
NM_203282.3 | NP_975011.3 | 2 | 220 | |||
XM_011528443.1 | XP_011526745.1 | 4 | 488 | |||
XM_011528444.1 | XP_011526746.1 | 5 | 508 | |||
XM_017027514.1 | XP_016883003.1 | 4 | 418 | |||
XM_017027515.1 | XP_016883004.1 | 6 | 845 | |||
XM_017027516.1 | XP_016883005.1 | 2 | 2892 | |||
XM_017027517.1 | XP_016883006.1 | 2 | 648 | |||
GenBank mRNA | AF054180.1 | - | 2 | 206 | ||
AK291286.1 | - | 2 | 187 | |||
BC043147.1 | - | 2 | 192 | 5296940 | ||
BC044906.1 | - | 1 | 1795 | |||
BC056243.1 | - | 2 | 196 | |||
BC139749.1 | - | 2 | 151 | 40115903 | ||
BX281205.1 | - | 3 | 223 | |||
DA101675.1 | - | 4 | 345 | |||
DB286139.1 | - | 3 | 319 |
Gene Symbol: LOC400682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528477.2 | XP_011526779.2 | 2 | 63 |
Gene Symbol: ZNF100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173531.3 | NP_775802.2 | 3 | 337 | ||
GenBank mRNA | AK126551.1 | - | 3 | 287 | ||
AK308798.1 | - | 3 | 258 | |||
BC031248.1 | - | 2 | 305 | |||
BC035579.1 | - | 3 | 249 | 5576923 | ||
DA241750.1 | - | 3 | 337 |
Gene Symbol: ZNF107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013746.2 | NP_001013768.1 | 3 | 405 | ||
NM_001282359.1 | NP_001269288.1 | 2 | 245 | |||
NM_001282360.1 | NP_001269289.1 | 2 | 245 | |||
NM_016220.4 | NP_057304.1 | 5 | 674 | |||
XM_017012283.1 | XP_016867772.1 | 5 | 830 | |||
XM_017012284.1 | XP_016867773.1 | 4 | 703 | |||
XM_017012285.1 | XP_016867774.1 | 6 | 972 | |||
XM_017012286.1 | XP_016867775.1 | 2 | 267 | |||
GenBank mRNA | AB027251.1 | - | 2 | 1215 | ||
AK303931.1 | - | 3 | 366 | |||
BC017809.1 | - | 5 | 624 | |||
BC047243.1 | - | 3 | 291 | 6066829 | ||
DA723595.1 | - | 2 | 245 |
Gene Symbol: ZNF208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007153.3 | NP_009084.2 | 2 | 194 | ||
XM_017027257.1 | XP_016882746.1 | 2 | 241 | |||
GenBank mRNA | AK096236.1 | - | 2 | 194 | ||
CB997649.1 | - | 2 | 221 |
Gene Symbol: ZNF253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021047.2 | NP_066385.2 | 2 | 137 | ||
GenBank mRNA | AF038951.1 | - | 1 | 45 | ||
AK074757.1 | - | 2 | 129 | |||
BC014148.2 | - | 2 | 137 | |||
BC125063.1 | - | 2 | 116 | 40117279 |
Gene Symbol: ZNF257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316996.1 | NP_001303925.1 | 3 | 351 | ||
NM_001316997.1 | NP_001303926.1 | 2 | 230 | |||
NM_033468.3 | NP_258429.2 | 2 | 230 | |||
GenBank mRNA | AF070651.1 | - | 2 | 131 | ||
AK056694.1 | - | 2 | 214 | |||
BC036446.1 | - | 3 | 345 | 4831021 | ||
BU155753.1 | - | 2 | 142 | |||
DB001158.1 | - | 2 | 194 |
Gene Symbol: ZNF273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021148.2 | NP_066971.2 | 2 | 220 | ||
NR_003099.1 | 3 | 309 | ||||
XM_011515746.2 | XP_011514048.1 | 2 | 251 | |||
XM_017011688.1 | XP_016867177.1 | 2 | 246 | |||
XM_017011689.1 | XP_016867178.1 | 7 | 6234 | |||
XM_017011690.1 | XP_016867179.1 | 6 | 6145 | |||
XM_017011691.1 | XP_016867180.1 | 7 | 3436 | |||
XM_017011692.1 | XP_016867181.1 | 5 | 5620 | |||
XM_017011693.1 | XP_016867182.1 | 6 | 1326 | |||
XM_017011694.1 | XP_016867183.1 | 6 | 684 | |||
XM_017011695.1 | XP_016867184.1 | 3 | 295 | |||
XM_017011696.1 | XP_016867185.1 | 4 | 521 | |||
XM_017011697.1 | XP_016867186.1 | 5 | 6056 | |||
XM_017011698.1 | XP_016867187.1 | 4 | 1173 | |||
GenBank mRNA | AK090648.1 | - | 6 | 1327 | ||
BC007078.1 | - | 3 | 245 | |||
BC008386.1 | - | 2 | 206 | |||
BC019234.1 | - | 3 | 249 | |||
BC063818.1 | - | 3 | 275 | 6503687 | ||
BC108740.1 | - | 3 | 240 | |||
DA324061.1 | - | 2 | 215 | |||
DA807335.1 | - | 3 | 309 |
Gene Symbol: ZNF429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001415.2 | NP_001001415.2 | 2 | 182 | ||
XM_017026748.1 | XP_016882237.1 | 3 | 681 | |||
XM_017026749.1 | XP_016882238.1 | 3 | 2595 | |||
XM_017026750.1 | XP_016882239.1 | 3 | 785 | |||
XM_017026751.1 | XP_016882240.1 | 2 | 696 | |||
XM_017026752.1 | XP_016882241.1 | 2 | 238 | |||
XM_017026753.1 | XP_016882242.1 | 4 | 534 | |||
XM_017026754.1 | XP_016882243.1 | 3 | 479 | |||
XM_017026755.1 | XP_016882244.1 | 4 | 2618 | |||
XM_017026756.1 | XP_016882245.1 | 3 | 486 | |||
GenBank mRNA | AF325191.1 | - | 2 | 158 | ||
AK308708.1 | - | 2 | 213 | |||
AY269786.1 | - | 2 | 158 |
Gene Symbol: ZNF486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052852.3 | NP_443084.2 | 2 | 189 | ||
GenBank mRNA | BC008936.2 | - | 2 | 173 | ||
BC027608.1 | - | 2 | 194 | |||
BC117268.1 | - | 2 | 129 | 40125819 |
Gene Symbol: ZNF492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020855.2 | NP_065906.1 | 2 | 193 | ||
GenBank mRNA | AB040906.1 | - | 2 | 80 | ||
BP871882.1 | - | 2 | 193 |
Gene Symbol: ZNF493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076678.2 | NP_001070146.1 | 2 | 191 | ||
NM_145326.2 | NP_663299.2 | 2 | 191 | |||
GenBank mRNA | AK092178.1 | - | 2 | 177 | ||
BC006408.1 | - | 2 | 154 | |||
BX648842.1 | - | 2 | 181 | |||
DA804007.1 | - | 2 | 191 |
Gene Symbol: ZNF506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099269.2 | NP_001092739.1 | 2 | 193 | ||
NM_001145404.1 | NP_001138876.1 | 2 | 193 | |||
GenBank mRNA | AK095575.1 | - | 2 | 193 | ||
AK122848.1 | - | 2 | 193 | |||
AL136548.1 | - | 2 | 151 | |||
BC018100.1 | - | 2 | 165 | |||
DA122515.1 | - | 2 | 193 |
Gene Symbol: ZNF626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076675.2 | NP_001070143.1 | 2 | 195 | ||
NM_145297.3 | NP_660340.1 | 2 | 195 | |||
GenBank mRNA | BC007116.1 | - | 2 | 174 | ||
BC107803.1 | - | 2 | 199 | |||
BC150651.1 | - | 2 | 106 | 9057022 | ||
CR749233.1 | - | 2 | 147 |
Gene Symbol: ZNF676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001411.2 | NP_001001411.2 | 1 | 267 | ||
GenBank mRNA | BX281304.1 | - | 1 | 267 |
Gene Symbol: ZNF679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153363.2 | NP_699194.2 | 3 | 350 | ||
XM_017011797.1 | XP_016867286.1 | 3 | 311 | |||
GenBank mRNA | BC033523.1 | - | 3 | 351 | 4821935 |
Gene Symbol: ZNF680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130022.1 | NP_001123494.1 | 2 | 245 | ||
NM_178558.4 | NP_848653.2 | 2 | 245 | |||
XM_006715959.2 | XP_006716022.1 | 3 | 521 | |||
XM_017012126.1 | XP_016867615.1 | 3 | 521 | |||
XM_017012127.1 | XP_016867616.1 | 3 | 521 | |||
XM_017012128.1 | XP_016867617.1 | 3 | 521 | |||
XM_017012129.1 | XP_016867618.1 | 2 | 253 | |||
GenBank mRNA | AK122937.1 | - | 2 | 206 | ||
AK131240.1 | - | 2 | 159 | |||
BC030700.1 | - | 2 | 229 | 4837196 | ||
DA669757.1 | - | 2 | 245 |
Gene Symbol: ZNF681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138286.2 | NP_612143.2 | 2 | 187 | ||
GenBank mRNA | AK122869.1 | - | 2 | 191 | ||
BC136346.1 | - | 2 | 66 | 9020333 |
Gene Symbol: ZNF708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021269.2 | NP_067092.2 | 2 | 243 | ||
XM_017027201.1 | XP_016882690.1 | 2 | 145 | |||
XM_017027204.1 | XP_016882693.1 | 2 | 60 | |||
GenBank mRNA | AK128184.1 | - | 2 | 243 | ||
AK131527.1 | - | 2 | 243 | |||
DA811084.1 | - | 2 | 273 |
Gene Symbol: ZNF726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244038.1 | NP_001230967.1 | 2 | 158 | ||
GenBank mRNA | AB593087.1 | - | 2 | 121 | ||
BC046415.1 | - | 2 | 130 | 4821872 | ||
BE783649.1 | - | 2 | 130 | |||
BX475164.1 | - | 2 | 166 | |||
CN359557.1 | - | 2 | 158 |
Gene Symbol: ZNF728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267716.1 | NP_001254645.1 | 2 | 223 | ||
GenBank mRNA | DA326735.1 | - | 2 | 223 | ||
DB076533.1 | - | 2 | 202 |
Gene Symbol: ZNF729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242680.1 | NP_001229609.1 | 2 | 148 | ||
XM_017026122.1 | XP_016881611.1 | 2 | 194 | |||
GenBank mRNA | BX956721.1 | - | 2 | 148 |
Gene Symbol: ZNF730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277403.1 | NP_001264332.1 | 2 | 244 | ||
XM_017026114.1 | XP_016881603.1 | 2 | 161 | |||
XM_017026115.1 | XP_016881604.1 | 2 | 321 | |||
GenBank mRNA | AK131472.1 | - | 2 | 244 |
Gene Symbol: ZNF737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159293.1 | NP_001152765.1 | 2 | 225 | ||
XM_005259697.3 | XP_005259754.1 | 2 | 214 | |||
XM_006722597.3 | XP_006722660.1 | 2 | 214 | |||
XM_011527598.2 | XP_011525900.1 | 2 | 214 | |||
XM_011527599.2 | XP_011525901.1 | 2 | 214 | |||
XM_017026116.1 | XP_016881605.1 | 3 | 312 | |||
GenBank mRNA | DA043454.1 | - | 2 | 225 |
Gene Symbol: ZNF90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007138.1 | NP_009069.1 | 2 | 184 | ||
GenBank mRNA | BC137211.1 | - | 2 | 111 | 9021208 | |
CN386341.1 | - | 2 | 111 | |||
M61870.1 | - | 2 | 166 |
Gene Symbol: ZNF93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031218.3 | NP_112495.2 | 2 | 221 | ||
GenBank mRNA | AK096342.1 | - | 2 | 178 | ||
BC070372.1 | - | 2 | 188 | |||
BC107581.1 | - | 2 | 192 | |||
BC136718.1 | - | 2 | 73 | 9020708 | ||
BC136719.1 | - | 2 | 73 | 9020709 | ||
BG257405.1 | - | 2 | 151 | |||
BM842045.1 | - | 2 | 193 |
Gene Symbol: ZNF98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098626.1 | NP_001092096.1 | 2 | 194 | ||
XM_011527705.1 | XP_011526007.1 | 2 | 153 | |||
XM_017026373.1 | XP_016881862.1 | 2 | 476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
