siRNA Details
Gene Symbol: DNAJB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153614.3 | NP_705842.2 | 6 | 1384 | ||
XM_005273984.3 | XP_005274041.1 | 5 | 749 | |||
XM_011545004.2 | XP_011543306.1 | 8 | 1635 | |||
XM_011545005.2 | XP_011543307.1 | 7 | 1475 | |||
XM_011545007.2 | XP_011543309.1 | 6 | 1160 | |||
XM_011545009.2 | XP_011543311.1 | 8 | 1309 | |||
XM_011545015.2 | XP_011543317.1 | 3 | 502 | |||
XM_017017675.1 | XP_016873164.1 | 7 | 905 | |||
XM_017017676.1 | XP_016873165.1 | 8 | 1485 | |||
XM_017017677.1 | XP_016873166.1 | 7 | 1354 | |||
XM_017017678.1 | XP_016873167.1 | 6 | 1165 | |||
GenBank mRNA | AF516185.1 | - | 3 | 1356 | ||
AK058095.1 | - | 3 | 1356 | |||
AK128589.1 | - | 8 | 3429 | |||
AY138810.2 | - | 6 | 1384 | |||
AY325766.1 | - | 6 | 677 | |||
BC127689.1 | - | 1 | 491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
