siRNA Details
Gene Symbol: DNAJB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153614.3 | NP_705842.2 | 8 | 1562 | ||
XM_005273984.3 | XP_005274041.1 | 7 | 927 | |||
XM_011545004.2 | XP_011543306.1 | 10 | 1813 | |||
XM_011545005.2 | XP_011543307.1 | 9 | 1653 | |||
XM_011545007.2 | XP_011543309.1 | 8 | 1338 | |||
XM_011545009.2 | XP_011543311.1 | 10 | 1487 | |||
XM_011545015.2 | XP_011543317.1 | 5 | 680 | |||
XM_017017675.1 | XP_016873164.1 | 9 | 1083 | |||
XM_017017676.1 | XP_016873165.1 | 10 | 1663 | |||
XM_017017677.1 | XP_016873166.1 | 9 | 1532 | |||
XM_017017678.1 | XP_016873167.1 | 8 | 1343 | |||
GenBank mRNA | AF516185.1 | - | 5 | 1534 | ||
AK058095.1 | - | 5 | 1534 | |||
AK128589.1 | - | 10 | 3607 | |||
AY138810.2 | - | 8 | 1562 | |||
AY325766.1 | - | 8 | 855 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
