siRNA Details
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164356.1 | NP_001157828.1 | 5 | 772 | ||
NM_015963.5 | NP_057047.4 | 6 | 2215 | |||
XM_005247016.4 | XP_005247073.3 | 6 | 2529 | |||
XM_011511291.2 | XP_011509593.1 | 6 | 2098 | |||
XM_017004256.1 | XP_016859745.1 | 6 | 2009 | |||
GenBank mRNA | AF258556.1 | - | 6 | 1967 | ||
AK225608.1 | - | 5 | 740 | |||
BC069235.1 | - | 6 | 2145 | 6645198 | ||
BC071896.1 | - | 5 | 1181 | |||
BC094822.1 | - | 5 | 1181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
