siRNA Details
Gene Symbol: CYP3A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000777.4 | NP_000768.1 | 1 | 144 | ||
NM_001190484.2 | NP_001177413.1 | 1 | 144 | |||
NM_001291829.1 | NP_001278758.1 | 1 | 144 | |||
NM_001291830.1 | NP_001278759.1 | 1 | 144 | |||
NR_033807.2 | 1 | 144 | ||||
XM_006715859.2 | XP_006715922.1 | 1 | 111 | |||
XM_011515843.1 | XP_011514145.1 | 1 | 111 | |||
GenBank mRNA | AK223008.1 | - | 1 | 114 | ||
AK296205.1 | - | 1 | 111 | |||
AK299002.1 | - | 1 | 114 | |||
AK309166.1 | - | 1 | 114 | |||
AK313813.1 | - | 1 | 111 | |||
BC033862.1 | - | 1 | 103 | 5188554 | ||
BX537676.1 | - | 1 | 117 | |||
BX648856.1 | - | 1 | 115 | |||
J04813.1 | - | 1 | 101 | |||
L26985.1 | - | 1 | 85 | |||
X90579.1 | - | 1 | 92 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
