siRNA Details
Gene Symbol: CYP3A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202855.2 | NP_001189784.1 | 1 | 145 | ||
NM_017460.5 | NP_059488.2 | 1 | 145 | |||
XM_017011780.1 | XP_016867269.1 | 1 | 143 | |||
GenBank mRNA | AK298451.1 | - | 1 | 138 | ||
AK298462.1 | - | 1 | 144 | |||
AK312967.1 | - | 1 | 138 | |||
AY606313.2 | - | 1 | 38 | |||
BC069418.1 | - | 1 | 88 | 7262313 | ||
BC101631.1 | - | 1 | 134 | 8069137 | ||
D00003.1 | - | 1 | 98 | |||
DA639071.1 | - | 1 | 223 | |||
DQ924960.1 | - | 1 | 38 | |||
J04449.1 | - | 1 | 106 | |||
M13785.1 | - | 1 | 98 | |||
M14096.1 | - | 1 | 106 | |||
M18907.1 | - | 1 | 128 | |||
X12387.1 | - | 1 | 107 |
Gene Symbol: CYP3A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000777.4 | NP_000768.1 | 1 | 168 | ||
NM_001190484.2 | NP_001177413.1 | 1 | 168 | |||
NM_001291829.1 | NP_001278758.1 | 1 | 168 | |||
NM_001291830.1 | NP_001278759.1 | 1 | 168 | |||
NR_033807.2 | 1 | 168 | ||||
XM_006715859.2 | XP_006715922.1 | 1 | 135 | |||
XM_011515843.1 | XP_011514145.1 | 1 | 135 | |||
GenBank mRNA | AK223008.1 | - | 1 | 138 | ||
AK296205.1 | - | 1 | 135 | |||
AK299002.1 | - | 1 | 138 | |||
AK309166.1 | - | 1 | 138 | |||
AK313813.1 | - | 1 | 135 | |||
BC033862.1 | - | 1 | 127 | 5188554 | ||
BX537676.1 | - | 1 | 141 | |||
BX648856.1 | - | 1 | 139 | |||
DC383127.1 | - | 1 | 169 | |||
J04813.1 | - | 1 | 125 | |||
L26985.1 | - | 1 | 109 | |||
X90579.1 | - | 1 | 116 |
Gene Symbol: CYP3A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000765.4 | NP_000756.3 | 1 | 145 | ||
GenBank mRNA | BC067436.1 | - | 1 | 123 | 7002135 | |
BX643367.1 | - | 1 | 142 | |||
D00408.1 | - | 1 | 41 |
Gene Symbol: CYP3A7-CYP3A51P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256497.2 | NP_001243426.2 | 1 | 145 | ||
GenBank mRNA | AF315325.1 | - | 1 | 38 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
