siRNA Details
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004965.6 | NP_004956.5 | 5 | 412 | ||
XM_011529543.1 | XP_011527845.1 | 8 | 1139 | |||
XM_011529544.1 | XP_011527846.1 | 7 | 1093 | |||
XM_011529545.1 | XP_011527847.1 | 7 | 473 | |||
XM_011529547.1 | XP_011527849.1 | 5 | 322 | |||
XM_011529549.1 | XP_011527851.1 | 7 | 634 | |||
XM_017028332.1 | XP_016883821.1 | 7 | 1923 | |||
XM_017028333.1 | XP_016883822.1 | 8 | 1995 | |||
XM_017028334.1 | XP_016883823.1 | 7 | 1905 | |||
XM_017028335.1 | XP_016883824.1 | 6 | 427 | |||
XM_017028336.1 | XP_016883825.1 | 7 | 556 | |||
XM_017028338.1 | XP_016883827.1 | 5 | 405 | |||
GenBank mRNA | AB209245.1 | - | 4 | 3365 | ||
AK056033.1 | - | 2 | 782 | |||
AK315410.1 | - | 5 | 355 | |||
BC000075.2 | - | 5 | 332 | 3510312 | ||
BC023984.1 | - | 5 | 324 | 4104558 | ||
BC070153.1 | - | 5 | 350 | 6450636 | ||
BC070154.1 | - | 5 | 330 | 6495327 | ||
BC071643.1 | - | 5 | 320 | 5549431 | ||
BC072672.1 | - | 5 | 321 | 6166317 | ||
BC106080.1 | - | 5 | 345 | 3855939 | ||
BC107741.1 | - | 5 | 418 | 6495117 | ||
BE779939.1 | - | 472 | ||||
BT007337.1 | - | 1 | 208 | |||
J02621.1 | - | 5 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
