siRNA Details
Gene Symbol: CLEC18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011880.2 | NP_001011880.2 | 9 | 1216 | ||
XM_005255976.2 | XP_005256033.1 | 9 | 1293 | |||
XM_011523093.2 | XP_011521395.1 | 8 | 999 | |||
XM_017023255.1 | XP_016878744.1 | 10 | 1470 | |||
XM_017023256.1 | XP_016878745.1 | 8 | 1186 | |||
GenBank mRNA | AY358373.1 | - | 9 | 1138 | ||
BM695392.1 | - | 5 | 427 |
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136214.2 | NP_001129686.1 | 10 | 1154 | ||
NM_001271197.1 | NP_001258126.1 | 10 | 1234 | |||
NM_182619.3 | NP_872425.2 | 10 | 1260 | |||
XM_005255934.1 | XP_005255991.1 | 9 | 1203 | |||
XM_017023206.1 | XP_016878695.1 | 9 | 1203 | |||
XM_017023207.1 | XP_016878696.1 | 8 | 1033 | |||
XM_017023208.1 | XP_016878697.1 | 9 | 1386 | |||
XM_017023209.1 | XP_016878698.1 | 8 | 1096 | |||
GenBank mRNA | AF521893.1 | - | 10 | 1273 | ||
AK074773.1 | - | 9 | 1088 | |||
AK289884.1 | - | 9 | 1202 | |||
BC078143.1 | - | 10 | 1136 | 5761001 |
Gene Symbol: CLEC18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173619.3 | NP_775890.2 | 9 | 1122 | ||
XM_006721179.1 | XP_006721242.1 | 9 | 1218 | |||
XM_011523034.1 | XP_011521336.1 | 10 | 1464 | |||
XM_011523036.2 | XP_011521338.1 | 8 | 1048 | |||
XM_011523037.1 | XP_011521339.1 | 8 | 1111 | |||
GenBank mRNA | AL833339.1 | - | 7 | 2220 | ||
BC039068.1 | - | 8 | 1904 | 5188643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
