siRNA Details
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136214.2 | NP_001129686.1 | 3 | 287 | ||
NM_001271197.1 | NP_001258126.1 | 3 | 367 | |||
NM_182619.3 | NP_872425.2 | 3 | 393 | |||
XM_005255934.1 | XP_005255991.1 | 2 | 336 | |||
XM_017023206.1 | XP_016878695.1 | 2 | 336 | |||
XM_017023207.1 | XP_016878696.1 | 2 | 406 | |||
XM_017023208.1 | XP_016878697.1 | 3 | 759 | |||
XM_017023209.1 | XP_016878698.1 | 2 | 336 | |||
GenBank mRNA | AF428259.1 | - | 2 | 406 | ||
AF521893.1 | - | 3 | 406 | |||
AK074773.1 | - | 2 | 221 | |||
AK096051.1 | - | 1 | 587 | |||
AK289884.1 | - | 2 | 335 | |||
AK298310.1 | - | 2 | 239 | |||
BC078143.1 | - | 3 | 269 | 5761001 | ||
BC141808.1 | - | 3 | 367 | 40147524 | ||
DC362064.1 | - | 3 | 393 |
Gene Symbol: CLEC18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011880.2 | NP_001011880.2 | 2 | 349 | ||
XM_005255976.2 | XP_005256033.1 | 2 | 426 | |||
XM_011523093.2 | XP_011521395.1 | 2 | 372 | |||
XM_017023255.1 | XP_016878744.1 | 3 | 603 | |||
XM_017023256.1 | XP_016878745.1 | 2 | 426 | |||
GenBank mRNA | AK293982.1 | - | 2 | 349 | ||
AY358373.1 | - | 2 | 271 | |||
DA375446.1 | - | 2 | 240 | |||
DC309733.1 | - | 2 | 349 |
Gene Symbol: CLEC18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173619.3 | NP_775890.2 | 2 | 255 | ||
XM_006721179.1 | XP_006721242.1 | 2 | 336 | |||
XM_011523034.1 | XP_011521336.1 | 3 | 582 | |||
XM_011523035.1 | XP_011521337.1 | 2 | 336 | |||
XM_011523036.2 | XP_011521338.1 | 2 | 421 | |||
XM_011523037.1 | XP_011521339.1 | 2 | 336 | |||
GenBank mRNA | AF448856.1 | - | 2 | 406 | ||
AL833339.1 | - | 2 | 342 | |||
BC039068.1 | - | 2 | 403 | 5188643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
