siRNA Details
Gene Symbol: LOC729737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039983.2 | 3 | 2648 |
Gene Symbol: LINC00999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024497.2 | 8 | 2052 | |||
GenBank mRNA | AK128746.1 | - | 1 | 609 |
Gene Symbol: LINC01000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024368.1 | 5 | 2536 | |||
GenBank mRNA | AK294679.1 | - | 1 | 264 |
Gene Symbol: LINC01001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028326.1 | 3 | 2325 |
Gene Symbol: LINC01002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028324.1 | 3 | 1145 | |||
GenBank mRNA | AK128780.1 | - | 1 | 684 | ||
AK303004.1 | - | 3 | 1109 |
Gene Symbol: LOC100132062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028325.1 | 3 | 2325 |
Gene Symbol: LOC100132287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028322.1 | 3 | 2325 | |||
GenBank mRNA | AK311622.1 | - | 1 | 258 |
Gene Symbol: LOC100133331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028327.1 | 3 | 2325 |
Gene Symbol: LOC101060022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544347.2 | XP_011542649.1 | 1 | 2752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
