siRNA Details
Gene Symbol: MAFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002359.3 | NP_002350.1 | 3 | 278 | ||
NM_032711.3 | NP_116100.2 | 3 | 310 | |||
XM_011523578.1 | XP_011521880.1 | 3 | 1213 | |||
GenBank mRNA | AF059195.1 | - | 3 | 262 | ||
BC012327.1 | - | 3 | 274 | 4637130 | ||
BI763651.1 | - | 4 | 280 | |||
DR156847.1 | - | 3 | 278 | |||
U84249.1 | - | 3 | 196 | |||
Y11514.1 | - | 3 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
