siRNA Details
Gene Symbol: NCOA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145260.1 | NP_001138732.1 | 9 | 1065 | ||
NM_001145261.1 | NP_001138733.1 | 9 | 1065 | |||
NM_001145262.1 | NP_001138734.1 | 8 | 994 | |||
NM_001145263.1 | NP_001138735.1 | 8 | 968 | |||
NM_005437.3 | NP_005428.1 | 8 | 908 | |||
GenBank mRNA | AK129911.1 | - | 8 | 848 | ||
AK292480.1 | - | 8 | 994 | |||
AK293978.1 | - | 6 | 625 | |||
AK302794.1 | - | 5 | 461 | |||
AK304123.1 | - | 9 | 966 | |||
AK310849.1 | - | 9 | 958 | |||
AK312325.1 | - | 8 | 914 | |||
AL162047.1 | - | 8 | 905 | |||
BC001562.1 | - | 8 | 849 | 3454306 | ||
BC012736.2 | - | 8 | 845 | 3842427 | ||
L49399.1 | - | 7 | 765 | |||
X77548.1 | - | 8 | 841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
