siRNA Details
Gene Symbol: PROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000313.3 | NP_000304.2 | 15 | 2228 | ||
NM_001314077.1 | NP_001301006.1 | 16 | 2315 | |||
GenBank mRNA | AK292994.1 | - | 15 | 2034 | ||
AK303895.1 | - | 8 | 1155 | |||
BC015801.1 | - | 15 | 2204 | 3909023 | ||
KU178283.1 | - | 15 | 1887 | |||
KU178284.1 | - | 16 | 1983 | |||
M14338.1 | - | 15 | 2055 | |||
M15036.1 | - | 15 | 2033 | |||
X12892.1 | - | 15 | 2055 | |||
Y00692.1 | - | 15 | 2010 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
