siRNA Details
Gene Symbol: EIF3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003754.2 | NP_003745.1 | 3 | 525 | ||
GenBank mRNA | AK027739.1 | - | 1 | 14 | ||
AK093511.1 | - | 5 | 1117 | |||
AK095574.1 | - | 4 | 487 | |||
AK289637.1 | - | 3 | 508 | |||
AK291354.1 | - | 3 | 505 | |||
AK293826.1 | - | 2 | 433 | |||
AK297621.1 | - | 4 | 358 | |||
BC000490.2 | - | 1 | 498 | 2819946 | ||
BC073753.1 | - | 3 | 493 | |||
BT006894.1 | - | 3 | 492 | |||
CD050033.1 | - | 3 | 525 | |||
CR456959.1 | - | 3 | 492 | |||
U94855.1 | - | 3 | 498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
