siRNA Details
Gene Symbol: ZDHHC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024630.2 | NP_078906.2 | 2 | 760 | ||
NM_153746.1 | NP_714968.1 | 2 | 760 | |||
XM_011536120.2 | XP_011534422.1 | 2 | 1341 | |||
XM_011536123.2 | XP_011534425.1 | 2 | 152 | |||
XM_017011307.1 | XP_016866796.1 | 2 | 445 | |||
XM_017011308.1 | XP_016866797.1 | 2 | 445 | |||
XM_017011309.1 | XP_016866798.1 | 2 | 445 | |||
XM_017011310.1 | XP_016866799.1 | 2 | 445 | |||
XM_017011311.1 | XP_016866800.1 | 2 | 445 | |||
XM_017011312.1 | XP_016866801.1 | 2 | 460 | |||
XM_017011313.1 | XP_016866802.1 | 2 | 2919 | |||
GenBank mRNA | AF542388.1 | - | 2 | 760 | ||
AF542389.1 | - | 2 | 760 | |||
AK309408.1 | - | 1 | 14 | |||
BC125072.1 | - | 3 | 277 | |||
BC125073.1 | - | 3 | 277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
