siRNA Details
Gene Symbol: DPY19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173812.4 | NP_776173.3 | 8, 9 | 1136 | ||
XM_006719352.2 | XP_006719415.1 | 8, 9 | 1062 | |||
XM_011538215.2 | XP_011536517.1 | 6, 7 | 602 | |||
XM_017019188.1 | XP_016874677.1 | 8, 9 | 1183 | |||
XM_017019189.1 | XP_016874678.1 | 8, 9 | 1183 | |||
XM_017019190.1 | XP_016874679.1 | 8, 9 | 1183 | |||
XM_017019193.1 | XP_016874682.1 | 7, 8 | 718 | |||
XM_017019194.1 | XP_016874683.1 | 7, 8 | 745 | |||
XM_017019195.1 | XP_016874684.1 | 8, 9 | 793 | |||
XM_017019196.1 | XP_016874685.1 | 8, 9 | 1183 | |||
XM_017019197.1 | XP_016874686.1 | 7, 8 | 1016 | |||
XM_017019198.1 | XP_016874687.1 | 8, 9 | 1183 | |||
XM_017019200.1 | XP_016874689.1 | 8, 9 | 1183 | |||
GenBank mRNA | AK057511.1 | - | 8, 9 | 1107 | ||
AL833344.1 | - | 8, 9 | 1063 | |||
AY358792.1 | - | 8, 9 | 1057 | |||
BC031225.1 | - | 2, 3 | 138 | |||
BC068442.1 | - | 8, 9 | 1139 | 5297524 | ||
BC125215.1 | - | 8, 9 | 1026 | |||
BC125216.1 | - | 8, 9 | 1026 |
Gene Symbol: DPY19L2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002833.2 | 8, 9 | 1189 | |||
GenBank mRNA | AK304236.1 | - | 8, 9 | 1349 | ||
BC066987.1 | - | 8, 9 | 1189 | |||
BX648666.1 | - | 3, 4 | 356 |
Gene Symbol: DPY19L2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003561.2 | 11, 12 | 2576 | |||
GenBank mRNA | AK093485.1 | - | 3, 4 | 203 | ||
AK131353.1 | - | 11, 12 | 2569 | |||
BC150309.1 | - | 8, 9 | 2183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
