siRNA Details
Gene Symbol: DPY19L2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003561.2 | 12 | 2655 | |||
NR_027768.1 | 10 | 2870 | ||||
GenBank mRNA | AK093485.1 | - | 4 | 282 | ||
AK131353.1 | - | 12 | 2648 | |||
AL834175.1 | - | 10 | 2623 | |||
BC150309.1 | - | 9 | 2262 |
Gene Symbol: DPY19L2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002833.2 | 9 | 1268 | |||
GenBank mRNA | AK304236.1 | - | 9 | 1428 | ||
BC066987.1 | - | 9 | 1268 | |||
BX648666.1 | - | 4 | 435 |
Gene Symbol: LOC107986726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012901.1 | XP_016868390.1 | 3 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
