siRNA Details
Gene Symbol: DPY19L2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002833.2 | 10 | 1315 | |||
GenBank mRNA | BC066987.1 | - | 10 | 1315 | ||
BX648666.1 | - | 5 | 482 |
Gene Symbol: DPY19L2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003561.2 | 13 | 2702 | |||
NR_027768.1 | 11 | 2917 | ||||
GenBank mRNA | AK093485.1 | - | 5 | 329 | ||
AK131353.1 | - | 13 | 2695 | |||
AL834175.1 | - | 11 | 2670 | |||
BC150309.1 | - | 10 | 2309 |
Gene Symbol: LOC107986726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012901.1 | XP_016868390.1 | 4 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
