siRNA Details
Gene Symbol: H3F3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005324.4 | NP_005315.1 | 2 | 307 | ||
GenBank mRNA | AF218029.1 | - | 2 | 131 | ||
AK130772.1 | - | 2 | 151 | |||
AK304607.1 | - | 2 | 151 | |||
AK312762.1 | - | 2 | 152 | |||
BC001124.2 | - | 2 | 135 | 2988279 | ||
BC006497.2 | - | 2 | 135 | 2959756 | ||
BC012813.2 | - | 2 | 135 | 2959756 | ||
BC017558.1 | - | 2 | 139 | 3910031 | ||
BC108701.1 | - | 2 | 114 | 6144255 | ||
BM723811.1 | - | 2 | 299 | |||
BX537379.1 | - | 2 | 152 | |||
BX647280.1 | - | 2 | 150 | |||
DB466880.1 | - | 2 | 306 |
Gene Symbol: H3F3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013699.2 | NP_001013721.2 | 1 | 93 | ||
GenBank mRNA | BC066906.1 | - | 1 | 93 | 5267388 | |
HQ873957.1 | - | 1 | 93 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
