siRNA Details
Gene Symbol: NPEPPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006310.3 | NP_006301.3 | 4 | 739 | ||
XM_011525496.1 | XP_011523798.1 | 5 | 541 | |||
XM_017025372.1 | XP_016880861.1 | 4 | 701 | |||
XM_017025373.1 | XP_016880862.1 | 5 | 541 | |||
GenBank mRNA | AJ132583.1 | - | 4 | 469 | ||
AK095877.1 | - | 5 | 505 | |||
AK096709.1 | - | 6 | 526 | |||
AK296887.1 | - | 4 | 490 | |||
AK297107.1 | - | 3 | 243 | |||
AK303037.1 | - | 5 | 534 | |||
AK311414.1 | - | 330 | ||||
BC065294.1 | - | 4 | 428 | 6059589 |
Gene Symbol: LOC101929950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722258.3 | XP_006722321.1 | 3 | 359 | ||
XM_017025500.1 | XP_016880989.1 | 3 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
