siRNA Details
Gene Symbol: NPEPPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006310.3 | NP_006301.3 | 3 | 609 | ||
XM_011525496.1 | XP_011523798.1 | 4 | 411 | |||
XM_017025372.1 | XP_016880861.1 | 3 | 571 | |||
XM_017025373.1 | XP_016880862.1 | 4 | 411 | |||
GenBank mRNA | AJ132583.1 | - | 3 | 339 | ||
AK095877.1 | - | 4 | 375 | |||
AK096709.1 | - | 5 | 396 | |||
AK296887.1 | - | 3 | 360 | |||
AK297107.1 | - | 2 | 113 | |||
AK303037.1 | - | 4 | 404 | |||
AK311414.1 | - | 84 | ||||
BC065294.1 | - | 3 | 298 | 6059589 | ||
DB476543.3 | - | 3 | 609 |
Gene Symbol: LOC101929950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722258.3 | XP_006722321.1 | 2 | 229 | ||
XM_017025500.1 | XP_016880989.1 | 2 | 234 |
Gene Symbol: LOC440434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036750.2 | 3 | 483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
