siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 3, 4 | 279 | ||
NR_003494.1 | 3, 4 | 279 | ||||
XM_006716262.1 | XP_006716325.1 | 3, 4 | 155 | |||
XM_011543842.2 | XP_011542144.1 | 3, 4 | 229 | |||
XM_017013916.1 | XP_016869405.1 | 3, 4 | 278 | |||
GenBank mRNA | AK098138.1 | - | 3, 4 | 246 | ||
AL529666.3 | - | 2, 3 | 216 | |||
BC062615.1 | - | 3, 4 | 324 | |||
BC085016.1 | - | 4, 5 | 352 | |||
BC098427.1 | - | 3, 4 | 329 | |||
BE729618.1 | - | 3, 4 | 221 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273603.4 | XP_005273660.1 | 3, 4 | 309 | ||
XM_005273607.2 | XP_005273664.1 | 3, 4 | 291 | |||
XM_017013740.1 | XP_016869229.1 | 3, 4 | 229 | |||
XM_017013741.1 | XP_016869230.1 | 2, 3 | 166 | |||
GenBank mRNA | AK291001.1 | - | 3, 4 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
