siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 2 | 177 | ||
NR_003494.1 | 2 | 177 | ||||
XM_006716256.3 | XP_006716319.1 | 2 | 170 | |||
XM_006716257.3 | XP_006716320.1 | 2 | 179 | |||
XM_006716260.3 | XP_006716323.1 | 2 | 167 | |||
XM_006716262.1 | XP_006716325.1 | 2 | 53 | |||
XM_011543842.2 | XP_011542144.1 | 2 | 127 | |||
XM_011543843.2 | XP_011542145.1 | 2 | 127 | |||
XM_017013916.1 | XP_016869405.1 | 2 | 176 | |||
XM_017013917.1 | XP_016869406.1 | 2 | 178 | |||
XM_017013918.1 | XP_016869407.1 | 2 | 150 | |||
XM_017013919.1 | XP_016869408.1 | 2 | 179 | |||
GenBank mRNA | AK308513.1 | - | 2 | 242 | ||
BC098427.1 | - | 2 | 227 | |||
BE729618.1 | - | 2 | 119 |
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 2 | 213 | ||
NM_201400.3 | NP_958802.1 | 2 | 213 | |||
NM_201598.3 | NP_963892.1 | 2 | 213 | |||
XM_005255157.4 | XP_005255214.1 | 2 | 186 | |||
XM_005255158.4 | XP_005255215.1 | 3 | 259 | |||
XM_011522404.2 | XP_011520706.1 | 3 | 259 | |||
GenBank mRNA | AY037162.1 | - | 2 | 181 | ||
BC010084.2 | - | 2 | 133 | 2822323 | ||
BG024981.1 | - | 2 | 181 | |||
BQ921410.1 | - | 2 | 196 | |||
CA495371.1 | - | 2 | 134 | |||
HY017433.1 | - | 2 | 213 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 2 | 132 | ||
XM_005273603.4 | XP_005273660.1 | 2 | 207 | |||
XM_005273604.4 | XP_005273661.1 | 2 | 207 | |||
XM_005273607.2 | XP_005273664.1 | 2 | 189 | |||
XM_006716387.3 | XP_006716450.1 | 2 | 207 | |||
XM_011544618.2 | XP_011542920.1 | 2 | 177 | |||
XM_011544622.2 | XP_011542924.1 | 2 | 127 | |||
XM_017013740.1 | XP_016869229.1 | 2 | 127 | |||
XM_017013742.1 | XP_016869231.1 | 2 | 127 | |||
XM_017013743.1 | XP_016869232.1 | 2 | 207 | |||
GenBank mRNA | AK297741.1 | - | 2 | 156 | ||
AK309984.1 | - | 2 | 185 |
Gene Symbol: FAM86B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024361.1 | 2 | 167 | |||
NR_024362.1 | 2 | 264 | ||||
NR_024363.1 | 2 | 264 | ||||
GenBank mRNA | AK307256.1 | - | 2 | 234 | ||
BC035792.1 | - | 2 | 142 |
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 2 | 158 | ||
NM_018172.2 | NP_060642.2 | 2 | 158 | |||
NM_152563.2 | NP_689776.1 | 2 | 158 | |||
GenBank mRNA | AK001523.1 | - | 2 | 132 | ||
AK130709.1 | - | 2 | 155 | |||
AK293221.1 | - | 2 | 132 | |||
AK311428.1 | - | 2 | 132 | |||
AK311518.1 | - | 2 | 155 | |||
AK311675.1 | - | 2 | 159 | |||
AK315454.1 | - | 2 | 132 | |||
BC032519.1 | - | 2 | 158 | 5494088 | ||
BQ921333.1 | - | 2 | 128 | |||
DC312827.1 | - | 2 | 158 |
Gene Symbol: FAM86C2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024249.1 | 2 | 155 |
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 2 | 234 | |||
GenBank mRNA | BC043342.2 | - | 2 | 60 | ||
BC068108.1 | - | 2 | 167 | |||
BC094682.1 | - | 2 | 281 | |||
DA753838.1 | - | 2 | 234 |
Gene Symbol: FAM86KP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513917.2 | XP_011512219.1 | 2 | 132 | ||
XM_017008895.1 | XP_016864384.1 | 2 | 132 | |||
XM_017008897.1 | XP_016864386.1 | 2 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
