siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 1, 2 | 151 | ||
NR_003494.1 | 1, 2 | 151 | ||||
XM_006716256.3 | XP_006716319.1 | 1, 2 | 144 | |||
XM_006716257.3 | XP_006716320.1 | 1, 2 | 153 | |||
XM_006716260.3 | XP_006716323.1 | 1, 2 | 141 | |||
XM_017013916.1 | XP_016869405.1 | 1, 2 | 150 | |||
XM_017013917.1 | XP_016869406.1 | 1, 2 | 152 | |||
XM_017013918.1 | XP_016869407.1 | 1, 2 | 124 | |||
XM_017013919.1 | XP_016869408.1 | 1, 2 | 153 | |||
GenBank mRNA | AK098138.1 | - | 1, 2 | 118 | ||
AK308439.1 | - | 1, 2 | 130 | |||
BC085016.1 | - | 2, 3 | 224 | |||
BE729618.1 | - | 1, 2 | 93 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 1, 2 | 106 | ||
XM_005273603.4 | XP_005273660.1 | 1, 2 | 181 | |||
XM_005273604.4 | XP_005273661.1 | 1, 2 | 181 | |||
XM_005273607.2 | XP_005273664.1 | 1, 2 | 163 | |||
XM_006716387.3 | XP_006716450.1 | 1, 2 | 181 | |||
XM_011544618.2 | XP_011542920.1 | 1, 2 | 151 | |||
XM_017013743.1 | XP_016869232.1 | 1, 2 | 181 | |||
GenBank mRNA | AK291001.1 | - | 1, 2 | 163 | ||
AK297741.1 | - | 1, 2 | 130 |
Gene Symbol: FAM86B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024361.1 | 1, 2 | 141 | |||
GenBank mRNA | BC035792.1 | - | 1, 2 | 116 |
Gene Symbol: FAM86KP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513917.2 | XP_011512219.1 | 1, 2 | 106 | ||
XM_017008895.1 | XP_016864384.1 | 1, 2 | 106 | |||
XM_017008897.1 | XP_016864386.1 | 1, 2 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
