siRNA Details
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 3 | 456 | |||
NR_033936.3 | 2 | 367 | ||||
NR_103496.2 | 2 | 317 | ||||
GenBank mRNA | AK309420.1 | - | 3 | 407 | ||
AK309732.1 | - | 2 | 318 | |||
AK310769.1 | - | 2 | 270 | |||
AK311124.1 | - | 2 | 222 | |||
BC065260.1 | - | 3 | 373 | |||
BC072390.1 | - | 2 | 320 | |||
DB070142.1 | - | 3 | 456 |
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102747.1 | 2 | 363 | |||
NR_102748.1 | 2 | 313 | ||||
GenBank mRNA | AK291684.1 | - | 3 | 357 | ||
AK307285.1 | - | 2 | 258 | |||
AK309746.1 | - | 2 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
