siRNA Details
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102748.1 | 2 | 378 | |||
GenBank mRNA | AK291684.1 | - | 3 | 422 | ||
AK307285.1 | - | 2 | 323 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 3 | 521 | |||
NR_033936.3 | 2 | 432 | ||||
NR_103496.2 | 2 | 382 | ||||
GenBank mRNA | AK026448.1 | - | 3 | 306 | ||
AK309732.1 | - | 2 | 383 | |||
AK310769.1 | - | 2 | 335 | |||
AK311124.1 | - | 2 | 287 | |||
AK311170.1 | - | 2 | 342 | |||
BC065260.1 | - | 3 | 438 | |||
BC072390.1 | - | 2 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
