siRNA Details
Gene Symbol: SCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080514.2 | NP_001073983.1 | 1 | 159 | ||
XM_006716616.2 | XP_006716679.1 | 2 | 1248 | |||
GenBank mRNA | CD672767.1 | - | 1 | 159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
