siRNA Details
Gene Symbol: RPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000970.4 | NP_000961.2 | 6 | 711 | ||
NM_001024662.2 | NP_001019833.1 | 6 | 887 | |||
NM_001320137.1 | NP_001307066.1 | 6 | 892 | |||
NM_001320138.1 | NP_001307067.1 | 6 | 926 | |||
NM_001320139.1 | NP_001307068.1 | 6 | 773 | |||
NM_001320140.1 | NP_001307069.1 | 6 | 935 | |||
NM_001320141.1 | NP_001307070.1 | 8 | 1000 | |||
NM_001320142.1 | NP_001307071.1 | 6 | 647 | |||
XM_017019781.1 | XP_016875270.1 | 8 | 2035 | |||
XM_017019782.1 | XP_016875271.1 | 8 | 1196 | |||
GenBank mRNA | AF261087.1 | - | 1 | 681 | ||
AK298117.1 | - | 4 | 413 | |||
AK299470.1 | - | 7 | 641 | |||
AY320410.1 | - | 3 | 231 | |||
BC004138.2 | - | 1 | 687 | 2823733 | ||
BC013863.1 | - | 1 | 661 | |||
BC020679.1 | - | 1 | 675 | 4616668 | ||
BC022444.1 | - | 6 | 842 | 4286423 | ||
BC031009.1 | - | 1 | 675 | 4333761 | ||
BC032299.1 | - | 1 | 672 | 4994660 | ||
BC071912.1 | - | 1 | 664 | 6293404 | ||
BC104824.1 | - | 6 | 687 | 8143827 | ||
BC104826.1 | - | 6 | 687 | 8143829 | ||
BG030827.1 | - | 1 | 437 | |||
CD049618.1 | - | 6 | 857 | |||
D17554.1 | - | 6 | 677 | |||
S71022.1 | - | 5 | 521 | |||
X69391.1 | - | 6 | 682 |
Gene Symbol: RPL6P27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126570.1 | - | 1423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
