siRNA Details
Gene Symbol: RPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000970.4 | NP_000961.2 | 7 | 829 | ||
NM_001024662.2 | NP_001019833.1 | 7 | 1005 | |||
NM_001320137.1 | NP_001307066.1 | 7 | 1010 | |||
NM_001320138.1 | NP_001307067.1 | 7 | 1044 | |||
NM_001320139.1 | NP_001307068.1 | 7 | 891 | |||
NM_001320140.1 | NP_001307069.1 | 7 | 1053 | |||
NM_001320141.1 | NP_001307070.1 | 9 | 1118 | |||
NM_001320142.1 | NP_001307071.1 | 7 | 765 | |||
XM_017019781.1 | XP_016875270.1 | 9 | 2153 | |||
XM_017019782.1 | XP_016875271.1 | 9 | 1314 | |||
GenBank mRNA | AF261087.1 | - | 1 | 799 | ||
AI583261.1 | - | 1 | 106 | |||
AK298117.1 | - | 5 | 531 | |||
AK299470.1 | - | 8 | 759 | |||
AK311863.1 | - | 7 | 793 | |||
AY320410.1 | - | 4 | 349 | |||
BC004138.2 | - | 1 | 805 | 2823733 | ||
BC013863.1 | - | 1 | 779 | |||
BC020679.1 | - | 1 | 793 | 4616668 | ||
BC022444.1 | - | 7 | 960 | 4286423 | ||
BC031009.1 | - | 1 | 793 | 4333761 | ||
BC032299.1 | - | 1 | 790 | 4994660 | ||
BC071912.1 | - | 1 | 782 | 6293404 | ||
BC104824.1 | - | 7 | 805 | 8143827 | ||
BC104826.1 | - | 7 | 805 | 8143829 | ||
BG030827.1 | - | 1 | 555 | |||
D17554.1 | - | 7 | 795 | |||
S71022.1 | - | 6 | 638 | |||
X69391.1 | - | 7 | 800 |
Gene Symbol: RPL6P27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126570.1 | - | 1541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
