siRNA Details
Gene Symbol: RPL6P27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126570.1 | - | 1446 |
Gene Symbol: RPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000970.4 | NP_000961.2 | 6 | 734 | ||
NM_001024662.2 | NP_001019833.1 | 6 | 910 | |||
NM_001320137.1 | NP_001307066.1 | 6 | 915 | |||
NM_001320138.1 | NP_001307067.1 | 6 | 949 | |||
NM_001320139.1 | NP_001307068.1 | 6 | 796 | |||
NM_001320140.1 | NP_001307069.1 | 6 | 958 | |||
NM_001320141.1 | NP_001307070.1 | 8 | 1023 | |||
NM_001320142.1 | NP_001307071.1 | 6 | 670 | |||
XM_017019781.1 | XP_016875270.1 | 8 | 2058 | |||
XM_017019782.1 | XP_016875271.1 | 8 | 1219 | |||
GenBank mRNA | AF261087.1 | - | 1 | 704 | ||
AK298117.1 | - | 4 | 436 | |||
AK299470.1 | - | 7 | 664 | |||
AK311863.1 | - | 6 | 698 | |||
AY320410.1 | - | 3 | 254 | |||
BC004138.2 | - | 1 | 710 | 2823733 | ||
BC013863.1 | - | 1 | 684 | |||
BC022444.1 | - | 6 | 865 | 4286423 | ||
BC031009.1 | - | 1 | 698 | 4333761 | ||
BC032299.1 | - | 1 | 695 | 4994660 | ||
BC071912.1 | - | 1 | 687 | 6293404 | ||
BC104824.1 | - | 6 | 710 | 8143827 | ||
BC104826.1 | - | 6 | 710 | 8143829 | ||
BG030827.1 | - | 1 | 460 | |||
D17554.1 | - | 6 | 700 | |||
S71022.1 | - | 5 | 544 | |||
X69391.1 | - | 6 | 705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
