siRNA Details
Gene Symbol: MEIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009813.2 | NP_001009813.1 | 9, 10 | 1328 | ||
NM_001301059.1 | NP_001287988.1 | 9, 10 | 1379 | |||
NM_020160.2 | NP_064545.1 | 9, 10 | 1517 | |||
XM_011527136.2 | XP_011525438.1 | 9, 10 | 1526 | |||
XM_011527138.2 | XP_011525440.1 | 7, 8 | 1376 | |||
XM_011527139.2 | XP_011525441.1 | 9, 10 | 1047 | |||
XM_011527140.1 | XP_011525442.1 | 9, 10 | 1546 | |||
XM_017027011.1 | XP_016882500.1 | 9, 10 | 1577 | |||
GenBank mRNA | AB209455.1 | - | 4, 5 | 557 | ||
AK098710.1 | - | 9, 10 | 1241 | |||
AK289950.1 | - | 9, 10 | 1111 | |||
AY091593.1 | - | 7, 8 | 608 | |||
BC025404.1 | - | 9, 10 | 1280 | 4825964 | ||
BC045741.1 | - | 2, 3 | 469 | |||
BC069251.1 | - | 9, 10 | 950 | 6211294 | ||
BI522513.1 | - | 2, 3 | 172 |
Gene Symbol: MEIS3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002211.1 | 1 | 650 | |||
GenBank mRNA | AK054814.1 | - | 1 | 649 | ||
U68385.1 | - | 1 | 432 |
Gene Symbol: MEIS3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289643.1 | - | 1 | 1116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
