siRNA Details
Gene Symbol: RPS3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006.4 | NP_000997.1 | 3, 4 | 437 | ||
GenBank mRNA | AK129825.1 | - | 3, 4 | 380 | ||
AK291075.1 | - | 3, 4 | 381 | |||
AK296121.1 | - | 3, 4 | 342 | |||
AK311788.1 | - | 3, 4 | 380 | |||
BC000204.1 | - | 1 | 369 | 3350750 | ||
BC001708.1 | - | 3, 4 | 399 | 3544072 | ||
BC004981.1 | - | 3, 4 | 369 | 2906117 | ||
BC006298.2 | - | 1 | 362 | 4079651 | ||
BC009219.2 | - | 1 | 369 | 3927639 | ||
BC009404.2 | - | 1 | 371 | 4308955 | ||
BC017123.2 | - | 1 | 369 | 3922810 | ||
BC018954.2 | - | 1 | 371 | |||
BC019072.2 | - | 1 | 420 | 5020581 | ||
BC030161.2 | - | 1 | 420 | 5020904 | ||
BC066926.1 | - | 1 | 381 | 4826149 | ||
BC070211.1 | - | 1 | 382 | 5809691 | ||
BC071916.1 | - | 1 | 369 | 6285242 | ||
BQ055367.1 | - | 3, 4 | 408 | |||
CX754149.1 | - | 1 | 437 | |||
L13802.1 | - | 3, 4 | 363 | |||
M77234.1 | - | 3, 4 | 374 | |||
M84711.1 | - | 3, 4 | 393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
