siRNA Details
Gene Symbol: TUBA3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080386.3 | NP_525125.2 | 3 | 540 | ||
GenBank mRNA | AK093116.1 | - | 3 | 454 | ||
AK292253.1 | - | 3 | 427 | |||
BC057810.1 | - | 3 | 426 | 5268074 | ||
DB051220.1 | - | 3 | 543 |
Gene Symbol: TUBA3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207312.2 | NP_997195.1 | 3 | 448 | ||
GenBank mRNA | AK131043.1 | - | 3 | 426 | ||
BC057811.1 | - | 3 | 448 | 5298532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
