siRNA Details
Gene Symbol: TUBA3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207312.2 | NP_997195.1 | 4 | 932 | ||
GenBank mRNA | AA397983.1 | - | 1 | 210 | ||
AK131043.1 | - | 4 | 910 | |||
BC057811.1 | - | 4 | 932 | 5298532 |
Gene Symbol: TUBA3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080386.3 | NP_525125.2 | 4 | 1024 | ||
GenBank mRNA | AK292253.1 | - | 4 | 911 | ||
AW966211.1 | - | 1 | 257 | |||
BC057810.1 | - | 4 | 910 | 5268074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
