siRNA Details
Gene Symbol: TUBA3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080386.3 | NP_525125.2 | 4 | 1096 | ||
GenBank mRNA | AK292253.1 | - | 4 | 983 | ||
AW966211.1 | - | 1 | 329 | |||
BC057810.1 | - | 4 | 982 | 5268074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
