siRNA Details
Gene Symbol: ATF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030287.3 | NP_001025458.1 | 3 | 439 | ||
NM_001040619.2 | NP_001035709.1 | 3 | 592 | |||
NM_001206484.2 | NP_001193413.2 | 3 | 552 | |||
NM_001206486.2 | NP_001193415.1 | 3 | 202 | |||
NM_001206488.2 | NP_001193417.2 | 3 | 569 | |||
NM_001674.3 | NP_001665.1 | 3 | 592 | |||
XM_005273146.1 | XP_005273203.1 | 3 | 801 | |||
XM_011509579.1 | XP_011507881.1 | 3 | 474 | |||
GenBank mRNA | AB066566.1 | - | 2 | 285 | ||
AB078026.1 | - | 2 | 285 | |||
AB078027.1 | - | 2 | 285 | |||
AB209032.1 | - | 1 | 3055 | |||
AK294296.1 | - | 3 | 461 | |||
AK312998.1 | - | 3 | 461 | |||
AL560585.3 | - | 3 | 591 | |||
AU135799.1 | - | 3 | 364 | |||
AY313926.1 | - | 2 | 285 | |||
AY313927.1 | - | 3 | 198 | |||
BC006322.2 | - | 3 | 450 | 4138076 | ||
BT006996.1 | - | 2 | 285 | |||
CD638624.1 | - | 3 | 369 | |||
CD641946.1 | - | 3 | 439 | |||
CR450334.1 | - | 2 | 285 | |||
DA589280.1 | - | 3 | 421 | |||
DA691940.1 | - | 3 | 418 | |||
DB197400.1 | - | 3 | 459 | |||
L19871.1 | - | 3 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
