siRNA Details
Gene Symbol: PNRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017761.3 | NP_060231.1 | 3 | 887 | ||
XM_017001691.1 | XP_016857180.1 | 3 | 792 | |||
GenBank mRNA | AF151042.1 | - | 3 | 871 | ||
AF374386.1 | - | 3 | 805 | |||
BC001959.1 | - | 3 | 858 | 3536977 | ||
BC078177.1 | - | 3 | 839 | 6526331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
