siRNA Details
Gene Symbol: RPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005.4 | NP_000996.2 | 5 | 495 | ||
NM_001256802.1 | NP_001243731.1 | 5 | 495 | |||
NM_001260506.1 | NP_001247435.1 | 5 | 543 | |||
NM_001260507.1 | NP_001247436.1 | 4 | 364 | |||
GenBank mRNA | AB062288.1 | - | 5 | 431 | ||
AK129794.1 | - | 5 | 467 | |||
AK130960.1 | - | 5 | 417 | |||
AK223048.1 | - | 5 | 444 | |||
AK313051.1 | - | 5 | 464 | |||
BC003137.1 | - | 5 | 444 | 2906158 | ||
BC003577.1 | - | 5 | 436 | |||
BC013196.1 | - | 5 | 436 | |||
BC013231.1 | - | 5 | 456 | |||
BC029981.1 | - | 4 | 389 | |||
BC034149.1 | - | 5 | 468 | 4665438 | ||
BC071917.1 | - | 5 | 447 | 6297012 | ||
BC100284.1 | - | 5 | 456 | 6160514 | ||
BC150501.1 | - | 3 | 270 | |||
BE797417.1 | - | 288 | ||||
BM831460.1 | - | 5 | 494 | |||
BM928330.1 | - | 5 | 525 | |||
BQ645330.1 | - | 503 | ||||
S42658.1 | - | 5 | 446 | |||
U14990.1 | - | 5 | 438 | |||
U14991.1 | - | 5 | 438 | |||
U14992.1 | - | 5 | 438 | |||
X55715.1 | - | 5 | 460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
