siRNA Details
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004327.3 | NP_004318.3 | 21 | 4123 | ||
NM_021574.2 | NP_067585.2 | 20 | 3991 | |||
GenBank mRNA | AB209991.1 | - | 21 | 3872 | ||
AK122842.1 | - | 4 | 1387 | |||
AK124310.1 | - | 1 | 1817 | |||
AK303241.1 | - | 21 | 2557 | |||
BC066122.1 | - | 21 | 3954 | |||
X02596.1 | - | 21 | 4015 | |||
Y00661.1 | - | 20 | 3991 |
Gene Symbol: BCRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037566.1 | 4 | 652 | |||
GenBank mRNA | AK124943.1 | - | 4 | 652 |
Gene Symbol: LOC102724751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030164.1 | XP_016885653.1 | 1 | 185 | ||
XM_017030165.1 | XP_016885654.1 | 1 | 185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
