siRNA Details
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004327.3 | NP_004318.3 | 22 | 4177 | ||
NM_021574.2 | NP_067585.2 | 21 | 4045 | |||
GenBank mRNA | AB209991.1 | - | 22 | 3926 | ||
AK122842.1 | - | 5 | 1441 | |||
AK124310.1 | - | 1 | 2349 | |||
AK303241.1 | - | 22 | 2611 | |||
BC066122.1 | - | 22 | 4008 | |||
X02596.1 | - | 22 | 4069 | |||
Y00661.1 | - | 21 | 4045 |
Gene Symbol: BCRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037566.1 | 5 | 706 | |||
GenBank mRNA | AK124943.1 | - | 5 | 706 |
Gene Symbol: LOC102724751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030164.1 | XP_016885653.1 | 2 | 239 | ||
XM_017030165.1 | XP_016885654.1 | 2 | 239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
