siRNA Details
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 3, 4 | 930 | |||
GenBank mRNA | BC045636.1 | - | 3, 4 | 933 | ||
BC068610.1 | - | 3, 4 | 933 | |||
BC110795.1 | - | 1 | 283 |
Gene Symbol: SPDYE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175064.2 | NP_778234.2 | 2, 3 | 633 | ||
XM_005249719.3 | XP_005249776.1 | 4, 5 | 1144 | |||
XM_017012015.1 | XP_016867504.1 | 4, 5 | 1082 | |||
XM_017012016.1 | XP_016867505.1 | 3, 4 | 494 | |||
XM_017012017.1 | XP_016867506.1 | 1, 2 | 729 | |||
GenBank mRNA | AF412027.1 | - | 2, 3 | 642 | ||
AL137266.1 | - | 2, 3 | 415 |
Gene Symbol: SPDYE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716210.3 | XP_006716273.1 | 2, 3 | 889 | ||
XM_017012933.1 | XP_016868422.1 | 2, 3 | 893 |
Gene Symbol: SPDYE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716211.3 | XP_006716274.1 | 2, 3 | 774 | ||
XM_017012887.1 | XP_016868376.1 | 2, 3 | 774 |
Gene Symbol: SPDYE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716212.3 | XP_006716275.1 | 2, 3 | 893 | ||
XM_017012888.1 | XP_016868377.1 | 2, 3 | 893 |
Gene Symbol: SPDYE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250742.4 | XP_005250799.1 | 3, 4 | 645 | ||
XM_005250743.4 | XP_005250800.1 | 2, 3 | 500 | |||
XM_011516756.2 | XP_011515058.1 | 2, 3 | 414 |
Gene Symbol: SPDYE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031618.3 | NP_001026789.2 | 4, 5 | 1094 | ||
XM_011516237.2 | XP_011514539.1 | 4, 5 | 1093 | |||
XM_017012223.1 | XP_016867712.1 | 4, 5 | 1093 | |||
GenBank mRNA | AK302217.1 | - | 4, 5 | 1094 | ||
BC100973.1 | - | 1, 2 | 332 | 40005226 | ||
DR731150.1 | - | 1, 2 | 238 | |||
JQ023160.1 | - | 3, 4 | 620 |
Gene Symbol: SPDYE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166339.1 | NP_001159811.1 | 4, 5 | 1094 | ||
XM_005250093.3 | XP_005250150.1 | 3, 4 | 1662 | |||
XM_011515702.2 | XP_011514004.1 | 3, 4 | 1662 |
Gene Symbol: SPDYE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004351.4 | NP_001004351.3 | 6, 7 | 1395 | ||
XM_011516234.2 | XP_011514536.1 | 6, 7 | 2298 | |||
XM_011516235.2 | XP_011514537.1 | 6, 7 | 2298 | |||
GenBank mRNA | BC056606.1 | - | 6, 7 | 1396 | 4838469 | |
BC100972.2 | - | 1, 2 | 320 |
Gene Symbol: SPDYE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306141.1 | NP_001293070.1 | 3, 4 | 1041 |
Gene Symbol: SPDYE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146210.2 | NP_001139682.2 | 3, 4 | 1041 |
Gene Symbol: SPDYE7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003666.2 | 1, 2 | 332 | |||
GenBank mRNA | BC100974.2 | - | 1, 2 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
