siRNA Details
Gene Symbol: SPDYE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031618.3 | NP_001026789.2 | 6 | 1208 | ||
XM_011516237.2 | XP_011514539.1 | 6 | 1207 | |||
XM_017012223.1 | XP_016867712.1 | 6 | 1207 | |||
GenBank mRNA | AK302217.1 | - | 6 | 1208 | ||
BC100973.1 | - | 3 | 446 | 40005226 | ||
DR731150.1 | - | 3 | 352 | |||
JQ023160.1 | - | 5 | 734 |
Gene Symbol: SPDYE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716210.3 | XP_006716273.1 | 4 | 1003 | ||
XM_017012933.1 | XP_016868422.1 | 4 | 1007 |
Gene Symbol: SPDYE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716211.3 | XP_006716274.1 | 4 | 888 | ||
XM_017012887.1 | XP_016868376.1 | 4 | 888 |
Gene Symbol: SPDYE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716212.3 | XP_006716275.1 | 4 | 1007 | ||
XM_017012888.1 | XP_016868377.1 | 4 | 1007 |
Gene Symbol: SPDYE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250742.4 | XP_005250799.1 | 5 | 759 | ||
XM_005250743.4 | XP_005250800.1 | 4 | 614 | |||
XM_011516756.2 | XP_011515058.1 | 4 | 528 |
Gene Symbol: SPDYE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166339.1 | NP_001159811.1 | 6 | 1208 | ||
XM_005250093.3 | XP_005250150.1 | 5 | 1776 | |||
XM_011515702.2 | XP_011514004.1 | 5 | 1776 |
Gene Symbol: SPDYE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004351.4 | NP_001004351.3 | 8 | 1509 | ||
XM_011516234.2 | XP_011514536.1 | 8 | 2412 | |||
XM_011516235.2 | XP_011514537.1 | 8 | 2412 | |||
GenBank mRNA | BC056606.1 | - | 8 | 1510 | 4838469 | |
BC100972.2 | - | 3 | 434 |
Gene Symbol: SPDYE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128076.1 | NP_001121548.1 | 4 | 641 | ||
XM_005256632.4 | XP_005256689.1 | 4 | 1510 | |||
XM_005256633.4 | XP_005256690.1 | 4 | 1510 | |||
GenBank mRNA | BC146949.1 | - | 4 | 641 | 9056877 | |
BC146966.1 | - | 4 | 641 | 9056894 |
Gene Symbol: SPDYE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306141.1 | NP_001293070.1 | 5 | 1155 |
Gene Symbol: SPDYE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146210.2 | NP_001139682.2 | 5 | 1155 |
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 5 | 1044 | |||
GenBank mRNA | BC045636.1 | - | 5 | 1047 | ||
BC068610.1 | - | 5 | 1047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
