siRNA Details
Gene Symbol: FEM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018708.2 | NP_061178.1 | 1 | 2093 | ||
GenBank mRNA | AJ311359.1 | - | 1 | 1953 | ||
AK074232.1 | - | 1 | 2051 | |||
AK126532.1 | - | 2 | 1821 | |||
AK315554.1 | - | 1 | 2065 | |||
AL359589.1 | - | 1 | 231 | |||
AL365408.1 | - | 1 | 548 | |||
AY356352.1 | - | 1 | 2047 | |||
BC004988.1 | - | 1 | 2051 | 2957950 | ||
CR749732.1 | - | 1 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
