siRNA Details
Gene Symbol: NUP50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007172.3 | NP_009103.2 | 6 | 1479 | ||
NM_153645.2 | NP_705931.1 | 7 | 1305 | |||
XM_005261312.1 | XP_005261369.1 | 7 | 1529 | |||
XM_005261314.1 | XP_005261371.1 | 7 | 1504 | |||
XM_006724103.1 | XP_006724166.1 | 6 | 1149 | |||
XM_006724104.1 | XP_006724167.1 | 7 | 1339 | |||
XM_011529833.1 | XP_011528135.1 | 7 | 1555 | |||
XM_017028537.1 | XP_016884026.1 | 7 | 1469 | |||
XM_017028538.1 | XP_016884027.1 | 5 | 1140 | |||
GenBank mRNA | AF107840.1 | - | 6 | 1182 | ||
AF116624.2 | - | 5 | 1034 | |||
AK095860.1 | - | 2 | 1216 | |||
AK304223.1 | - | 5 | 1138 | |||
AK314454.1 | - | 6 | 1338 | |||
BC016055.1 | - | 7 | 1414 | 4856954 | ||
BC028125.1 | - | 7 | 1305 | 5166132 | ||
BC070133.1 | - | 6 | 1339 | 6463796 | ||
CR456533.1 | - | 5 | 1033 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
